missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KV4.1 (KCND1) Polyclonal Antibody

Description
Immunogen sequence: NTLDRYPDTL LGSSEKEFFY DADSGEYFFD RDPDMFRHVL NFYRTGRLHC PRQECIQAFD EELAFYGLVP ELVGDCCLEE YRDRKKENAE RLAEDEEAEQ Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The KV gene family encodes more than 30 genes that comprise the subunits of the K+ channels, and they vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha-subunits (KV alpha), which include the KV1, KV2, KV3 and KV4 proteins, and accessory or KV beta subunits that modify the gating properties of the coexpressed KV alpha subunits. Differences exist in the patterns of trafficking, biosynthetic processing and surface expression of the major KV1 subunits (KV1. 1, KV1. 2 and KV1. 4) expressed in rat and human brain, suggesting that the individual protein subunits are highly regulated to control for the assembly and formation of functional neuronal channels.
Specifications
Specifications
| Antigen | KV4.1 (KCND1) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Kcnd1 |
| Gene Accession No. | Q9NSA2 |
| Gene Alias | 1110037K09Rik; Kca2-1; Kcnd1; KV4.1; mShal; mShal1; potassium channel, voltage gated Shal related subfamily D, member 1; potassium channel, voltage gated Shal-related subfamily D, member 1; potassium voltage gated channel Shal-related family member 1; potassium voltage gated channel, Shal-related family, member 1; potassium voltage-gated channel subfamily D member 1; potassium voltage-gated channel, Shal-related family, member 1; potassium voltage-gated channel, Shal-related subfamily, member 1; Shal; Shal1; Shal-type potassium channel; Voltage-gated potassium channel subunit Kv4.1 |
| Gene Symbols | Kcnd1 |
| Show More |