missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT71 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149902
Description
KRT71 Polyclonal Antibody for Western Blot, ICC/IF, IHC
K6IRS1 belongs to a family of type II keratins that are specifically expressed in the inner root sheath of hair follicles.Specifications
| KRT71 | |
| Polyclonal | |
| Unconjugated | |
| KRT71 | |
| CK-71; cytokeratin-71; hK6irs; hK6irs1; HYPT13; K6IRS1; K71; KB34; Keratin 6 irs; keratin 71; keratin 71, type II; keratin, type II cytoskeletal 71; Keratin-71; KRT6IRS; KRT6IRS1; KRT71; Type II inner root sheath-specific keratin-K6irs1; type-II keratin Kb34 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 112802, 56735, 683613 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q3SY84, Q9R0H5 | |
| KRT71 | |
| Recombinant Keratin 71 (KRT71) (Arg132-Glu440). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |