Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Replaceable Frit and Grownd Joint (90mm)
Recommended for large volume filtration analysis of corrosive liquids or samples with high particulate loads
$6380.18 - $6554.86
Specifications
Type Filtration Assembly
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
K953840-2090
View Documents
DWK Life Sciences
9538402090
2 L
Each for $6,380.18
Only null left
K9538404090
View Documents
DWK Life Sciences
9538404090
4 L
Case for $6,554.86
Only null left
Description
Description
  • The glass support disc can be replaced if it becomes clogged or if sample-to-sample contamination is a concern
  • Like the apparatus with fritted glass support, the ground joint connection eliminates the possibility of phthalate contamination that can occur when using silicone or neoprene stoppers
  • Each apparatus is supplied with 1000mL funnel, 90mm glass support base, 40-60 micron porosity fritted glass disc, anodized aluminum clamp and filtration flask
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Filtration Assembly
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'