Learn More
DWK Life Sciences KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (90mm)
Catalog No. K9538350090
Change view
Click to view available options
Volume (Metric):
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
K9538350090 2 L
K9538250090 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K9538350090 Supplier DWK Life Sciences Supplier No. 9538350090
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (90mm) is designed for large volume filtration analysis of corrosive liquids or samples with high particulate loads that would tend to clog a 47mm filter.
  • Designed for large volume filtration analysis of corrosive liquids or samples with high particulate loads that would tend to clog a 47 mm diameter filter
  • Also recommended for filtering large volumes of HPLC solvents
  • Ground glass connection eliminates the possibility of phthalate contamination that can occur when using silicone or neoprene stoppers
  • Each apparatus supplied with 1000 mL funnel, 90 mm 40 to 60 micron porosity fritted glass support base, anodized aluminum clamp, and filtration flask
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Disposable Reusable
Material Glass
Sterility Non-sterile
Type Microfiltration Assembly
Volume (Metric) 2 L
For Use With (Equipment) Filtration Assembly and vacuum pump