Learn More
DWK Life Sciences Kimble™ Kontes™ Standard Threaded Microscale Kit
Features versatile threaded chaste connections which can be assembled rightside up or upside down
Supplier:  DWK Life Sciences 7470000000
Catalog No. K747000-0000
Only null left
Description
Description
Contains all components necessary to perform experiments described in the following texts: Organic Laboratory Techniques: A Microscale Approach, Pavia, Lampman, Kriz, Engel, Saunders College Publishing: and Microscale Organic Laboratory, Mayo, Pike, Butcher, John Wiley and Sons
  • Fits easily into lab bench drawers
  • Also available in EKONO-CASE™, a low-cost corrugated cardboard alternative to plastic With high temperature resistant blue nylon/PTFE connectors
  • Ideal for use to 200°C
Order Info
Kits are packed in a rugged polyethylene storage case (10D x 13.5W x 2.5 in.H) with a die cut foam insert. Kit components are listed in the chart below. Contact your Fisher Scientific Customer Service Representative to order replacement items. Items without a catalog number in the chart are not available separately.
Specifications
Specifications
USP Type I and ASTM E438 Type I, Class A
13-425 claisen adapter, 13-425 connecting adapter, 13-425, 20-400 vacuum adapter, 13-425 to 0.25 in. - 28 connector, 13-425 to hose connector, 10ml micro beaker, 13-425 to 13-425 connector, 13-425 to 20-400 connector, 20-400 to 20-400 connector
Threaded Research Organic Chemistry Kit
Product Suggestions
Product Suggestions