Learn More
DWK Life Sciences Kimble™ Kontes™ Screw Thread Culture Tubes with Unattached Rubber Lined Caps
Uniform wall thickness for maximum heat transfer and heat resistance
$954.68 - $2,296.75
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
12141356
|
DWK Life Sciences
KC4506613100 |
8 mL |
Case of 144 Each for $1,428.39
|
|
|||||
|
01183006
|
DWK Life Sciences
KC4506616125 |
16 mL |
Case of 144 Each for $2,296.75
|
|
|||||
|
11417138
|
DWK Life Sciences
KC4506620150 |
30 mL |
Case of 96 Each for $1,863.18
|
|
|||||
|
12141359
|
DWK Life Sciences
KC4506625150 |
50 mL |
Case of 48 Each for $954.68
|
|
|||||
Description
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- KG-33 borosilicate glass
- Designed from ASTM™ E982, Type VI, Class A requirements
- Thread finish complies with Glass Packaging Institute (GPI) requirements
- Cap is not attached