Learn More
DWK Life Sciences Kimble™ Kontes™ Screw Thread Culture Tubes with Unattached Rubber Lined Caps
Uniform wall thickness for maximum heat transfer and heat resistance
Supplier:  DWK Life Sciences KC4506620150
 View more versions of this product
Catalog No. 11417138
Only null left
Description
Description
  • The 20 x 125 mm size of these tubes is suitable for the determination of insulin according to the method of Alving, Rubin and Millor, Journal of Biological Chemistry, 127, 3 (March 1939)
  • Provided with phenolic screw-thread caps, unattached
  • Series 45066 is designed from ASTM E982, Type VI, Class A requirements
  • Series 45066A is designed from ASTM E982, Type VI, Class B requirements
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Designed from ASTM E982, Type VI, Class A requirements
150 mm
18-415
Rubber Lined Screw Cap
30 mL
Borosilicate Glass (3.3)
20 mm
Product Suggestions
Product Suggestions