missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ PTFE Stirrer Blade

Stirrer blade for K786500 Series 10mm shafts

Supplier:  DWK Life Sciences 7890300024

 View more versions of this product

Catalog No. K7890300024


Only null left

Description

Description

  • Blades are 0.12 in. (3mm) thick
  • Use K789030-0022 Blades with K612000 and K612200 Series reaction flasks

Specifications

Chemical Identifiers

Case of 1
0.12 in.
0.3 cm

Specifications

5000mL Flask, 10mm Shaft
2.4 cm
35/25
0.3 cm
13 cm
0.9 in.
Case of 1
0.12 in.
5.1 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions