Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE-Plug Stopcocks, Graduations, Pressure Equalizing Lines, Ground Joint Funnels
Graduations to stopcock are numbered both up and down
Supplier:  DWK Life Sciences 6345300250
 View more versions of this product
Catalog No. K6345300250
Only null left
Description
Description
Without stoppers.
Drip tips extend below the standard taper joint to ensure delivery
Specifications
Chemical Identifiers
Borosilicate Glass
PTFE closure
Pressure equalizing lines, lower drip tip, stopcock
Addition Funnel
USP, ASTM E438, Type I, Class A
Yes
24/40
Specifications
250 mL
345 mm
PTFE closure
Pressure equalizing lines, lower drip tip, stopcock
24/40
Addition Funnel
Borosilicate Glass
USP, ASTM E438, Type I, Class A
Yes
5mL
2
Product Suggestions
Product Suggestions