missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ NMR Sample Tube Caps

Catalog No. K8970950011
Change view
Click to view available options
For Use With (Equipment):
10 mm NMR Sample Tube
5 mm NMR Sample Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
K8970950011 5 mm NMR Sample Tube
K8970951011 10 mm NMR Sample Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K8970950011 Supplier DWK Life Sciences Supplier No. 8970950011

For Kontes™ 5 or 10mm NMR sample tubes

  • Convenient 100-cap variety pack of tight-fitting polyethylene color caps
  • Stay in place even with a buildup of internal vaporization pressure

Order Info

Variety pack for 5mm (0.19 in. ) tubes includes 20 caps each of red, black, white, green and yellow. Variety pack for 10mm (0.39 in. ) tubes contains 25 each of red, black, green, and yellow.

Specifications

Includes 20 caps each of red, black, white, green and yellow
Product Type Pressure Cap Variety Pack
Material Polyethylene
Color Assorted
For Use With (Application) Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures
For Use With (Equipment) 5 mm NMR Sample Tube