Learn More
Kontes™ Membrane Filtration Glassware from Restek DFS Item
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
Supplier:  Chromatographic RKKT9538270000
 View more versions of this product
Catalog No. 06717526
Only null left
Explore available promotions
Description
Description
  • Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
  • Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
Specifications
Specifications
Microfiltration Apparatus
Glass
Flask (1 L)
1 L
Product Suggestions
Product Suggestions