Learn More
Kontes™ Membrane Filtration Glassware from Restek DFS Item
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
Supplier:  Chromatographic RKKT9537610000
 View more versions of this product
Catalog No. 06717521
Only null left
Explore available promotions
Description
Description
  • Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
  • Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
Specifications
Specifications
Microfiltration Apparatus
Funnel (100 mL)
47 mm
Glass
100 mL
Product Suggestions
Product Suggestions