Learn More
Kontes™ Membrane Filtration Glassware from Restek DFS Item
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
Supplier:  Chromatographic RKKT6760014035
 View more versions of this product
Catalog No. 06717518
Only null left
Explore available promotions
Description
Description
  • Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
  • Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
Specifications
Specifications
Microfiltration Apparatus
PTFE
40/35
Joint Sleeve
Product Suggestions
Product Suggestions