missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Heavy-Duty Serum Bottles

Catalog No. 1031018
Change view
Click to view available options
Capacity (Metric):
4000 mL
9000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
1031018 4000 mL
1031019 9000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1031018 Supplier DWK Life Sciences Supplier No. 149604

The heavy duty construction of these bottles is designed to prolong life expectancy with harder than normal usage. Ideal for packaging and storage.

  • Rounded shoulders
  • Neck is tooled for a uniform fit with a No.8 rubber stopper
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

Specifications

Capacity 135.2 oz., 4000mL
Capacity (Metric) 4000 mL
Capacity (English) 135.2 oz.
Diameter (English) 6.57 in.
Material Borosilicate Glass
Height (Metric) 310 mm
Product Type Serum Bottle
Height (English) 12.2 in.
Closure Material Rubber
Closure Size No. 8
Diameter (Metric) 167 mm
Show More Show Less