Learn More
DWK Life Sciences Kimble™ Kontes™ Flasks with standard taper Necks
Single neck
Supplier: DWK Life Sciences 6010000429
Description
- Round bottom
Specifications
| 3.2 in. | |
| Borosilicate Glass | |
| Round |
| 250 mL | |
| 29/42 | |
| 83 mm |