Learn More
DWK Life Sciences Kimble™ Kontes™ Disposaflex™ Stopcocks
For Disposaflex™ Chromatography Columns
$425.79 - $931.00
Specifications
Type Stopcock
For Use With Disposaflex Chromatography Column
Material Polycarbonate
View More Specs
Products 2
Catalog Number Mfr. No. Port Type Price Quantity  
Catalog Number Mfr. No. Port Type Price Quantity  
K420163-0001
View Documents
DWK Life Sciences
4201630001
Luerlock F-Luer to M-Luer Lock
Pack of 10 Each for $425.79
Only null left
K420163-0000
View Documents
DWK Life Sciences
4201630000
1-Way F-Luer to M-Luer
Pack of 50 Each for $931.00
Only null left
Description
Description
For Disposaflex™ Chromatography Columns
  • Polycarbonate Body
  • HDPE Plug
Specifications
Specifications
Stopcock
Disposaflex Chromatography Column
Polycarbonate
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'