Learn More
DWK Life Sciences Kimble™ Kontes™ Disposaflex™ Stopcocks
For Disposaflex™ Chromatography Columns
Supplier:  DWK Life Sciences 4201630000
 View more versions of this product
Catalog No. K420163-0000
Only null left
Explore available promotions
Description
Description
For Disposaflex™ Chromatography Columns
  • Polycarbonate Body
  • HDPE Plug
Specifications
Specifications
Stopcock
1-Way F-Luer to M-Luer
Disposaflex Chromatography Column
Polycarbonate
Product Suggestions
Product Suggestions