missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kojic Acid Dipalmitate 98.0+%, TCI America™
$46.60 - $130.48
Chemical Identifiers
| CAS | 79725-98-7 |
|---|---|
| Molecular Formula | C38H66O6 |
| MDL Number | MFCD03840587 |
| Synonym | 5-(Palmitoyloxy)-2-[(palmitoyloxy)methyl]-4H-pyran-4-one |
Chemical Identifiers
| 79725-98-7 | |
| C38H66O6 | |
| MFCD03840587 | |
| 5-(Palmitoyloxy)-2-[(palmitoyloxy)methyl]-4H-pyran-4-one |
Specifications
| 79725-98-7 | |
| White | |
| MFCD03840587 | |
| 5-(Palmitoyloxy)-2-[(palmitoyloxy)methyl]-4H-pyran-4-one | |
| ≥98.0% (HPLC) | |
| Kojic Acid Dipalmitate |
| 96°C | |
| C38H66O6 | |
| 5 g | |
| 618.94 | |
| Crystalline Powder |
Safety and Handling
TSCA : No