accessibility menu, dialog, popup

UserName

CAS RN 79725-98-7

H3COOOOOOCH3

CAS RN 79725-98-7

IUPAC Name: 6-(hexadecanoyloxy)-4-oxo-4H-pyran-3-yl hexadecanoate
Synonyms: 6-(hexadecanoyloxy)-4-oxopyran-3-yl hexadecanoate
Molecular Weight (g/mol): 604.91
Molecular Formula: C37H64O6
InChi Key: URJOWNUVTORLNY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC1=CC(=O)C(OC(=O)CCCCCCCCCCCCCCC)=CO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)