missing translation for 'onlineSavingsMsg'
Learn More

Kojic Acid Dipalmitate 98.0+%, TCI America™

Catalog No. K00505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00505G 5 g
K005025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00505G Supplier TCI America Supplier No. K00505G
Only null left

Chemical Identifiers

CAS 79725-98-7
Molecular Formula C38H66O6
MDL Number MFCD03840587
Synonym 5-(Palmitoyloxy)-2-[(palmitoyloxy)methyl]-4H-pyran-4-one

Specifications

Melting Point 96°C
Color White
Quantity 5 g
Formula Weight 618.94
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Kojic Acid Dipalmitate