missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kojic Acid, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor K1062100GM
Specifications
| 501-30-4 | |
| 0.006 | |
| C6H6O4 | |
| BEJNERDRQOWKJM-UHFFFAOYSA-N | |
| 5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one | |
| 0.99 |
| 1 | |
| Amber Glass Bottle | |
| 100 g | |
| OCC1=CC(=O)C(O)=CO1 | |
| 142.11 | |
| Reagent |
Chemical Identifiers
| 501-30-4 | |
| 142.11 | |
| 5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one |
| C6H6O4 | |
| BEJNERDRQOWKJM-UHFFFAOYSA-N | |
| OCC1=CC(=O)C(O)=CO1 |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.