missing translation for 'onlineSavingsMsg'
Learn More

Kojic Acid, 99%, Spectrum™ Chemical

Catalog No. p-7165769
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18611163 25 g Amber Glass Bottle
18611162 100 g Amber Glass Bottle
18611161 1 kg Amber Glass Bottle
18611166 5 kg Poly Pail
18611164 25 kg Fiber Drum
18611167 50 kg Fiber Drum
6 options

Catalog No. 18611163

Supplier: Spectrum Chemical Mfg Cor K106225GM

Only null left

Chemical Identifiers

CAS 501-30-4
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
InChI Key BEJNERDRQOWKJM-UHFFFAOYSA-N
IUPAC Name 5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
SMILES OCC1=CC(=O)C(O)=CO1

Specifications

CAS Max % 1
Loss on Drying 0.006
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.