missing translation for 'onlineSavingsMsg'
Learn More

Kojic acid, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014344
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014344 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014344 Supplier Merck Emd Millipore Supplier No. 95197100MG
Only null left
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 501-30-4
Physical Form Neat
Linear Formula C6H6O4
MDL Number MFCD00006580
Melting Point 152°C to 155°C
Synonym 2-Hydroxymethyl-5-hydroxy-gamma-pyrone; 5-Hydroxy-2-hydroxymethyl-4 H-4-pyranone
Type Analytical Standard
Quantity 100 mg
Formula Weight 142.11
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Show More Show Less