missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Knife Switch

Knife Switch on a durable plastic base. Rugged construction that stands up to tough student use. Poles and knives are made of brass for good electrical connections.

$17.11 - $32.71

Specifications

Product Type Knife Switch
Material Plastic
For Use With (Application) For good electrical connections.
View More Specs

Products 3
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
S23687
View Documents Promotion Details
Eisco™
PH1000A
Single pole, Single throw
Each for $17.11
Only null left
 
S23688
View Documents Promotion Details
Eisco™
PH1000B
Single pole, Double throw
Each for $18.36
Only null left
 
S23690
View Documents Promotion Details
Eisco™
PH1002B
Double pole, Double throw
Each for $32.71
Only null left
 
Description

Description

  • Durable plastic base knife switch
  • Brass poles and knives for good electrical connections
Specifications

Specifications

Knife Switch
Plastic
For good electrical connections.
Videos
SDS
Documents

Documents