Learn More
KNF™ Neuberger 316SS Vacuum Pumps and Compressors
Designed for gas transfer applications (except two-stage) and for use with rotary evaporators, dryers, distillers, filtration devices, and as a general vacuum source for the lab
Supplier: KNF™ Neuberger 070496075643
Description
Pump corrosive gases without the need for cold traps, drinking water or pump oil. Eliminate the need to dispose of aspirator-generated wastewater or spent pump oil.
- Maximum pressure, 60psig (two-stage model is vacuum only)
- Type 316 stainless-steel and PTFE wetted parts
- Easily disassemble without the need for special tools
- Pump thread size is 1/4 NPTF
Order Info
For all 115V/60Hz. 230V/50Hz units is available; contact your Fisher Scientific™ Customer Service Representative to order.
Specifications
| 3.75 A | |
| Stainless Steel (Pump Head) | |
| Rotary evaporator, dryer, distiller, filtration device | |
| 10 in. | |
| 11 in. | |
| 10 in. | |
| 115 V |
| 115 V 60 Hz | |
| 30 L/min. | |
| 60 Hz | |
| 25 cm | |
| 28 cm | |
| 60 psi |