Learn More
Fisherbrand™ Knee Space Panels for Drawer Aprons
Fisherbrand™ Knee Space Panels for Drawer Aprons are to be used with Drawer aprons. The knee space panel closes off the casework to the wall to create a clear sitting or standing space under the work surface.
Description
Compliance
SEFA 8
Specifications
| 36 in. | |
| 2 in. | |
| 5.1 cm | |
| 23.4 in. | |
| Steel |
| Knee Space Panel | |
| 60 cm | |
| 91.4 cm | |
| White |