missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL24 Polyclonal Antibody

Description
This antibody is predicted to react with mouse and rat based on sequence homology.
KLHL24 is specifically reduces kainate receptor-mediated currents in hippocampal neurons, most probably by modulating channel properties.
Specifications
Specifications
| Antigen | KLHL24 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | KLHL24 |
| Gene Accession No. | Q6TFL4 |
| Gene Alias | 1110046J11Rik; 4930429H24Rik; 6530402O20Rik; C85082; DRE1; kainate receptor interacting protein for GluR6; kainate receptor-interacting protein for GluR6; kelch like family member 24; kelch-like 24; kelch-like family member 24; kelch-like protein 24; Klhl24; KRIP6; Protein DRE1 |
| Gene Symbols | KLHL24 |
| Show More |