missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KLF10 Recombinant Rabbit Monoclonal Antibody (23GB6605)
GREENER_CHOICE

Catalog No. MA553074
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553074 100 μL
MA553075 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553074 Supplier Invitrogen™ Supplier No. MA553074
Only null left

Rabbit Recombinant Monoclonal Antibody

KLF10 Recombinant Monoclonal Antibody for Western Blot

KLF10 is a nuclear protein belonging to the Sp1 C2H2-type zinc-finger protein family with three C2H2-type zinc fingers. KLF10 is a transcriptional repressor involved in the regulation of cell growth and inhibits cell growth. It binds to the consensus sequence 5'-GGTGTG-3' and is induced by TGF-beta thus playing a role in linking TGF-beta-mediated signaling cascades to the regulation of pancreatic epithelial cell growth. KLF10 is involved in inducing and promoting apoptosis through the mitochondrial apoptotic pathway and may inhibit Smad7 expression in bronchial epithelial cells. Ubiquitous expression of KLF10 is seen in most of the tissues. Reports suggest that it may mediate up-regulation of TGFBI in VHL-deficient tumors and other cancers. Over expression of KLF10 may be associated with renal clear cell carcinoma and other tumors. KLF11 is a zinc finger transcription factor that binds to SP1-like sequences in epsilon- and gamma-globin gene promoters. This binding inhibits cell growth and causes apoptosis. Defects in this gene are a cause of maturity-onset diabetes of the young type 7 (MODY7). Three transcript variants encoding two different isoforms have been found for this gene.

Specifications

Antigen KLF10
Applications Western Blot
Classification Recombinant Monoclonal
Clone 23GB6605
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KLF10
Gene Accession No. O08876, O89091, Q13118
Gene Alias AI115143; Cpg20; early growth response-alpha; EGR[a]; EGRA; Egral; EGR-alpha; Gdnfif; GDNF-inducible factor; Klf10; Krueppel-like factor 10; Kruppel like factor 10; Kruppel-like factor 10; mGIF; TGFB inducible early growth response; TGFB inducible early growth response 1; TGFB-inducible early growth response protein 1; Tieg; TIEG1; TIEG-1; transcription factor GIF; Transforming growth factor-beta-inducible early growth response protein 1; zinc finger transcription factor homolog CPG20; zinc finger transcription factor TIEG
Gene Symbols KLF10
Host Species Rabbit
Immunogen A synthesized peptide derived from human KLF10 (40-100AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21847, 7071, 81813
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less