missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KLC1 Recombinant Rabbit Monoclonal Antibody (23GB5925)
GREENER_CHOICE

Catalog No. MA552952
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552952 100 μL
MA552953 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552952 Supplier Invitrogen™ Supplier No. MA552952
Only null left

Rabbit Recombinant Monoclonal Antibody

KLC1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Conventional kinesin is a tetrameric molecule composed of two heavy chains and two light chains, and transports various cargos along microtubules toward their plus ends. The heavy chains provide the motor activity, while the light chains bind to various cargos. This gene encodes a member of the kinesin light chain family. It associates with kinesin heavy chain through an N-terminal domain, and six tetratricopeptide repeat (TPR) motifs are thought to be involved in binding of cargos such as vesicles, mitochondria, and the Golgi complex. Thus, kinesin light chains function as adapter molecules and not motors per se. Although previously named 'kinesin 2', this gene is not a member of the kinesin-2 / kinesin heavy chain subfamily of kinesin motor proteins. Extensive alternative splicing produces isoforms with different C-termini that are proposed to bind to different cargos; however, the full-length nature of some of these variants has not been determined.

Specifications

Antigen KLC1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5925
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KLC1
Gene Accession No. O88447, P37285, Q07866
Gene Alias AI874768; kinesin 2; kinesin 2 60/70kDa; kinesin light chain 1; Kinesin2; Klc; KLC 1; Klc1; KNS 2; KNS 2A; Kns2; KNS2A; medulloblastoma antigen MU-MB-2.50; MGC15245
Gene Symbols KLC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human KLC1 (501-573AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16593, 171041, 3831
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less