Learn More
BUCHI KjelMaster K-375/KjelSampler 376 System
Catalog No. 05405675
Change view
Click to view available options
Capacity:
1 Sample
24 samples
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
05405675 24 samples
05405674 1 Sample
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05405675 Supplier BUCHI Supplier No. 113751710
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
An automatic distillation unit designed for convenient nitrogen and protein analysis. The reduced need for user interaction, and intelligent software saves time and costs.
  • Compliant and flexible plug-and-measure titration concept (colorimetric, potentiometric)
  • Comprises a steam generator, automatic aspiration of residues, glass splash protector, and integrated titration
  • Fully automated dilution of the sample, alkali addition, receiver dispensing, potentiometric titration, and calculation/reporting
  • Safe operation, user protection and guided routine operation
  • Simple LIMS integration and traceable data recording
Specifications
Electrical Requirements 220/240 V, 50/60 Hz
Voltage 220/240 V
Includes Color Touchscreen (8.4 in.)
For Use With (Application) Kjeldahl Determination
Amperage 15 A
Type Kjeldahl Instrument
Capacity 24 samples