missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Kjeldahl Tablets
Description
- Environmentally friendly: free of toxic mercury and selenium
- Different tablets to fit individual needs
- For digestion
Specifications
Specifications
| Chemical Name or Material | Copper Micro |
| Melting Point | 1067°C (Main Component) |
| Quantity | 1000 Pk. |
| Packaging | Plastic Container |
| Color | Blue |
| CAS | 7758-99-8, 7778-80-5 |
| Molecular Formula | K2SO4; CuSO4 |
| Molecular Weight (g/mol) | 1.65 |
| Percent Purity | Mixture |
| Grade | Analytical |