missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kjeldahl Digestion Solution, Reagents
Specifications
Specifications
| Chemical Name or Material | Kjeldahl Digestion Solution |
| Boiling Point | 102°C |
| Color | Colorless |
| CAS | 7778-80-5, 7664-93-9, 7732-18-5 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | Poly Bottle |
| Synonym | Nitrogen |
| Grade | Reagent |