missing translation for 'onlineSavingsMsg'
Learn More

Kjeldahl Digestion Reagent with Copper Sulfate, APHA 4500-N(org) B, Reagents

Catalog No. p-200021675
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334856 1 L
01334855 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334856 Supplier Labchem Inc Supplier No. CK2004001A
Only null left

Kjeldahl Digestion Reagent with Copper Sulfate, APHA 4500-N(org) B, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 102°C
Color Blue
CAS 7778-80-5, 7664-93-9, 7732-18-5
Physical Form Liquid
Quantity 1 L
Packaging Poly Bottle
Grade Reagent
Chemical Name or Material Kjeldahl Digestion Reagent with Copper Sulfate, APHA 4500-N(org) B