missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kjel-Sorb™ Solution (Certified), Fisher Chemical™
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Specifications
| Absorbance | Pass Test |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Color | Red |
| Physical Form | Liquid |
| CAS | 10043-35-3 |
| Quantity | 20 L |
| Packaging | PolyPac™ |
| Grade | Certified |
| Chemical Name or Material | Kjel-Sorb™ Solution |
Safety and Handling
| DOTInformation | DOT Class 9, : Miscellaneous |