missing translation for 'onlineSavingsMsg'
Learn More

Kjel-Sorb™ Solution (Certified), Fisher Chemical™

Catalog No. SK154
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle, EcoSafPak™
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SK154 4 L Poly Bottle, EcoSafPak™
SK1520 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SK154 Supplier Thermo Fisher Scientific Supplier No. SK15 4
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Saturated boric acid solution with indicator for use in Kjeldahl determinations

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Absorbance Pass Test
Melting Point 0°C
Boiling Point 100°C
Color Red
Physical Form Liquid
CAS 10043-35-3
Quantity 4 L
Packaging Poly Bottle, EcoSafPak™
Grade Certified
Chemical Name or Material Kjel-Sorb™ Solution
Safety handling Check Alert storage code Red
DOTInformation DOT Class 9, : Miscellaneous