missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kirenol 98.0+%, TCI America™
$350.44 - $1049.46
Chemical Identifiers
| CAS | 52659-56-0 |
|---|---|
| Molecular Formula | C20H34O4 |
| Molecular Weight (g/mol) | 338.488 |
| MDL Number | MFCD00210519 |
| InChI Key | NRYNTARIOIRWAB-TWQVXVJFSA-N |
| PubChem CID | 129316850 |
| IUPAC Name | 1-[(2S,4aR,4bS,8R,8aR)-6-hydroxy-8-(hydroxymethyl)-2,4b,8-trimethyl-4,4a,5,6,7,8a,9,10-octahydro-3H-phenanthren-2-yl]ethane-1,2-diol |
| SMILES | CC1(CCC2C(=C1)CCC3C2(CC(CC3(C)CO)O)C)C(CO)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
K005925MG
|
TCI America
K005925MG |
25 mg |
Each for $350.44
|
|
|||||
|
K0059100MG
|
TCI America
K0059100MG |
100 mg |
Each for $1,049.46
|
|
|||||
Chemical Identifiers
| 52659-56-0 | |
| 338.488 | |
| NRYNTARIOIRWAB-TWQVXVJFSA-N | |
| 1-[(2S,4aR,4bS,8R,8aR)-6-hydroxy-8-(hydroxymethyl)-2,4b,8-trimethyl-4,4a,5,6,7,8a,9,10-octahydro-3H-phenanthren-2-yl]ethane-1,2-diol |
| C20H34O4 | |
| MFCD00210519 | |
| 129316850 | |
| CC1(CCC2C(=C1)CCC3C2(CC(CC3(C)CO)O)C)C(CO)O |
Specifications
| 52659-56-0 | |
| White-Yellow | |
| MFCD00210519 | |
| NRYNTARIOIRWAB-TWQVXVJFSA-N | |
| 1-[(2S,4aR,4bS,8R,8aR)-6-hydroxy-8-(hydroxymethyl)-2,4b,8-trimethyl-4,4a,5,6,7,8a,9,10-octahydro-3H-phenanthren-2-yl]ethane-1,2-diol | |
| 129316850 | |
| ≥98.0% (GC) | |
| Kirenol |
| 199°C | |
| C20H34O4 | |
| 25 mg | |
| CC1(CCC2C(=C1)CCC3C2(CC(CC3(C)CO)O)C)C(CO)O | |
| 338.488 | |
| 338.49 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : SF7352310
TSCA : No
Recommended Storage : Refrigerator