accessibility menu, dialog, popup

UserName

CAS RN 52659-56-0

CH3CH3HOHOCH3OHHO

CAS RN 52659-56-0

IUPAC Name: 1-[6-hydroxy-8-(hydroxymethyl)-2,4b,8-trimethyl-2,3,4,4a,4b,5,6,7,8,8a,9,10-dodecahydrophenanthren-2-yl]ethane-1,2-diol
Synonyms: 1-[6-hydroxy-8-(hydroxymethyl)-2,4b,8-trimethyl-4,4a,5,6,7,8a,9,10-octahydro-3H-phenanthren-2-yl]ethane-1,2-diol
Molecular Weight (g/mol): 338.49
Molecular Formula: C20H34O4
InChi Key: NRYNTARIOIRWAB-UHFFFAOYNA-N
SMILES: CC1(CO)CC(O)CC2(C)C3CCC(C)(C=C3CCC12)C(O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results