missing translation for 'onlineSavingsMsg'
Learn More

Kirenol 98.0+%, TCI America™

Catalog No. K005925MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K005925MG 25 mg
K0059100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K005925MG Supplier TCI America Supplier No. K005925MG
Only null left

Chemical Identifiers

CAS 52659-56-0
Molecular Formula C20H34O4
Molecular Weight (g/mol) 338.488
MDL Number MFCD00210519
InChI Key NRYNTARIOIRWAB-TWQVXVJFSA-N
PubChem CID 129316850
IUPAC Name 1-[(2S,4aR,4bS,8R,8aR)-6-hydroxy-8-(hydroxymethyl)-2,4b,8-trimethyl-4,4a,5,6,7,8a,9,10-octahydro-3H-phenanthren-2-yl]ethane-1,2-diol
SMILES CC1(CCC2C(=C1)CCC3C2(CC(CC3(C)CO)O)C)C(CO)O

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 25 mg
Formula Weight 338.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Kirenol
RTECSNumber SF7352310
TSCA No
Recommended Storage Refrigerator