missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kininogen Heavy Chain Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149901
Description
Kininogen Heavy Chain Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.Specifications
| Kininogen Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| KNG1 | |
| Alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; fitzgerald factor; High molecular weight kininogen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; KNG; KNG1; Low molecular weight growth-promoting factor; Lysyl-bradykinin; T-kinin; Williams-Fitzgerald-Flaujeac factor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 288001 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08934 | |
| KNG1 | |
| Recombinant High Molecular Weight Kininogen (HMWK) (Leu214-Asp475). | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody |