Learn More
Thermo Scientific™ KingFisher™ PlasmidPro Maxi Processor
The KingFisher™ PlasmidPro Maxi Processor transforms the maxi-scale plasmid purification process with a cutting-edge, fully automated workflow that eliminates labor-intensive centrifugation and pipetting steps.
Supplier: Thermo Scientific™ PPMX1000
Description
The KingFisher PlasmidPro Maxi Processor transforms the maxi-scale plasmid purification process with a cutting-edge, fully automated workflow that eliminates labor-intensive centrifugation and pipetting steps. Simply pour in the culture and let the instrument handle the rest. In under 75 minutes, this innovative system efficiently executes all essential purification steps, delivering high-quality, endotoxin-free plasmid DNA.
True walkaway automation
The KingFisher PlasmidPro system is fully automated, eliminating the need for pipetting or centrifugation steps. It completes each run in under 75 minutes, allowing you to focus on other tasks.
Exceptional quality
Optimized for maxi-scale plasmid DNA purification from 150 mL of bacterial culture, the system consistently delivers high-quality, advanced, transfection-grade plasmid DNA.
Convenient pre-filled cartridge
The one-piece, pre-filled KingFisher PlasmidPro Maxi Processor Endotoxin-Free Cartridge eliminates setup time and ensures a simple clean-up process, enabling quick turnaround for your next purification.
Effortless installation and setup
The installation process can be completed in minutes on any benchtop, ensuring immediate usability.
Optional AB Assurance Plan
The AB Assurance Plan is a comprehensive repair plan that aims to optimize instrument performance and ensure the availability of critical systems. It includes preventive maintenance, proactive instrument monitoring, and repairs, all aimed at keeping your laboratory operating smoothly.
Additional details
The KingFisher PlasmidPro Maxi Processor is specifically optimized for cultures grown in Luria-Bertani (LB) medium.
At the end of the procedure, the system delivers endotoxin-free plasmid DNA resuspended in TE buffer, ensuring optimal quality and compatibility with downstream applications.
Specifications
| Plasmid Prep | |
| Advanced Transfection Applications | |
| None | |
| 1 Sample/Run | |
| 7 in. Touchscreen | |
| USB | |
| 1 Ea. | |
| 1 Sample/Run | |
| Room temperature | |
| 100/240 VAC, 50/60 Hz (nominal), automatic voltage detection |
| Bacterial Culture | |
| 150 mL | |
| Cartridge | |
| 19.7 x 11.8 x 23.6 in. (50 x 30 x 60 cm) | |
| Plasmid purification, advanced transfection applications | |
| Plasmid DNA | |
| <75 min. | |
| 150 mL | |
| ~30 GB | |
| 50/60 Hz |
For Research Use Only. Not for use in diagnostic procedures.