missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ mL Purification System

Catalog No. 22387002
Encompass
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22387002 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22387002 Supplier Thermo Scientific™ Supplier No. 5400050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Automate low-throughput sample preparation in your laboratory workflow using Thermo Scientific™ KingFisher mL Purification System.

The KingFisher™ mL Purification System is discontinued. Thermo Fisher Scientific offers other automation platforms such as the Thermo Scientific™ KingFisher™ Duo Prime Purification System, KingFisher™ Flex Purification System and KingFisher Apex. The company will continue to support current KingFisher™ mL users with plastics, service, and technical support through 2028, or best effort.

Thermo Fisher Scientific has transitioned to limited support stage. It will offer limited support contracts and provide best-effort service and support, subject to availability of spare parts and resources.

If you are interested in upgrading to one of its newer sample preparation instruments, please contact your local account manager for product information.

Features:
  • Offers rapid and reproducible purification of high-quality DNA, RNA, proteins and cells for various types of downstream applications.
  • Open and flexible system lets customer use any magnetic particle-based kit to meet application demands
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification
  • Ready-made protocols for different type of applications available

Recommended for:

Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays

Specifications

Volume (Metric) Processing 50 to 1000 μL
Capacity 15 Samples/Run
Description KingFisher mL Purification System, 100-240V, 50/60Hz nominal
Collection Efficiency of Particles 95%
Display START/STOP/TWO CURSOR KEYS/LCD
For Use With (Application) Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays
Magnetic Rods 3 x 5 format
Particle Size (Metric) ca. >1 μm
Quantity Each
Tip Combo Special design, 1 x 5 format
Vessel Type Special tube strip, 1 x 5 tubes
Depth (English) 11.4 in.
Depth (Metric) 29 cm
Height (English) 12.2 in.
Height (Metric) 31 cm
Weight (English) 23 lb.
Weight (Metric) 10 kg
Width (English) 11.4 in.
Width (Metric) 29 cm
Electrical Requirements 100-240 V, 50/60 Hz
Show More Show Less
Do you have scripts for using the MagMAX kits on the KingFisher mL instrument?

We do not have prewritten scripts for using MagMAX kits on the KingFisher mL instrument. We recommend adapting scripts written for the KingFisher Flex or KingFisher Presto instruments. You will still need to validate the scripts on your KingFisher mL instrument.

Can I use the MagMAX Express-96 Deep Well Head (Cat. No. 4388435) on the KingFisher 96 instrument?

Yes, the MagMAX Express-96 Deep Well Head can be used on the KingFisher 96 instrument. However, it is not compatible with the KingFisher Flex instrument.

What should I do if the tip comb does not load correctly onto my Thermo Scientific KingFisher system?

If the tip does not load correctly onto a Thermo Scientific KingFisher magnetic head, we recommend that you try manually flexing the tips both lengthwise and widthwise to level the tip comb. A video of flexing the tip comb can be found here: https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/automated-purification-extraction/kingfisher-systems/resources.html. The video is under "KingFisher Flex" and titled "How to install the magnet head and check the tip comb." Flexing of the tip comb is demonstrated at timepoint 1:35. If that does not work, try a different box or lot of tips. This applies to all KingFisher instruments.

For research use only. Not for use in diagnostic procedures.