missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Purification System

Catalog No. 22387001
Encompass
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22387001 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22387001 Supplier Thermo Scientific™ Supplier No. 5400000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Automate time-consuming sample preparation processes of nucleic acids, proteins and cells with Thermo Scientific™ KingFisher™ Purification Systems.

The KingFisher™ Purification System is pending discontinuation. Thermo Fisher Scientific offers other automation platforms such as the KingFisher Duo Prime Purification System, KingFisher Flex Purification System and KingFisher Apex Purification System. The company will continue to support current KingFisher Purification System users with plastics, service, and technical support through 2028, or best effort.

By August 1st, 2023, Thermo Fisher Scientific will transition to limited support stage. It will offer limited support contracts and provide best-effort service and support, subject to availability of spare parts and resources.

If you are interested in upgrading to one of its newer sample preparation instruments, please contact your local account manager for product information.

Features:
  • Offer rapid and reproducible purification of high-quality DNA, RNA, proteins and cells for various types of downstream applications
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification
  • Ready-made protocols for different types of applications available

Recommended for:

Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays

Specifications

Volume (Metric) Processing 20 to 200 μL
Capacity 24 Samples/Run
Description Kingfisher Purification System, 100-240V, 50/60Hz
Collection Efficiency of Particles 99%
Display START/STOP/TWO CURSOR KEYS/LCD
For Use With (Application) Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays
Magnetic Rods 2 x 12 format
Particle Size (Metric) ca. >1 μm
Quantity Each
Tip Combo Special design, 2 x 12 format
Plate Type Special design, 8 x 12 grid format
Depth (English) 11.4 in.
Depth (Metric) 29 cm
Height (English) 12.2 in.
Height (Metric) 31 cm
Weight (English) 23 lb.
Weight (Metric) 10 kg
Width (English) 11.4 in.
Width (Metric) 29 cm
Electrical Requirements 100-240 V, 50/60 Hz
Show More Show Less
What is BindIt Software?

BindIt Software is used to operate the KingFisher 96, KingFisher mL, KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto instruments. With the software you can:
• Create a plate layout
• Create new or modify existing protocols
• Transfer protocols to the KingFisher instrument memory
• Delete protocols from the KingFisher instrument
• Execute a protocol directly from the software without first sending it to the KingFisher instrument memory
• Export protocols to other databases
• Import protocols to the PC
• View protocol status reports

For research use only. Not for use in diagnostic procedures.