missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Flex Purification System, SmartStart, 24 deep-well head

Catalog No. A39968
Encompass
Change view
Click to view available options
Quantity:
1 package
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39968 1 package
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39968 Supplier Thermo Scientific™ Supplier No. A39968

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Efficiently prepare samples from a variety of materials with the Thermo Scientific KingFisher Flex Purification System which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, or cell purification from virtually any source. Package includes the KingFisher Flex Purification System and SmartStart Orientation.

  • Fully automated system yields high-speed purification of nucleic acids, proteins, and cells
  • High-throughput system processes up to 96 samples in less than 20 minutes
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands
  • Easy-to-use BindIt Software provides instrument control, protocol creation, and modification
  • Ready-made protocols available for different type of applications
  • 1-day SmartStart Orientation to get \up and running quickly

Get up and running fast with SmartStart Orientation

  • The SmartStart Orientation included in this package is performed by a Field Application Scientist (FAS) in your lab and helps to -
    • Get up and running quickly
    • Get connected to your local network and Cloud
    • Choose appropriate reagents and consumables to meet specific needs
  • Content will vary depending on customer's level of knowledge and experience

Specifications

Volume (Metric) Processing 50 to 1000 μL
Plates per Deck 24
Capacity 24 Samples
Description KingFisher 24 DW plate
Collection Efficiency of Particles >95%
Display START / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / 4 cursor keys / LCD
For Use With (Application) Genomics and Proteomics, Target Identification, Veterinary Assays, Biomarker Discovery, Quality Control, Forensics
For Use With (Equipment) Nucleic Acid Purification Systems
Heating Temperature Ambient +5°C to 115°C
Magnetic Rods 24 in. one frame; interchangeable magnet heads
Particle Size (Metric) ca. >1 μm
Quantity 1 package
Plate Type 24 or 96-well plates, KingFisher 24 Deep Well plate (200 to 5000 μL), Microtiter 96 Deep Well plate (50 to 1000 μL), KingFisher 96 Plates (20 to 200 μL), PCR plate (20 to 100 μL)
Depth (English) 23.6 in.
Depth (Metric) 60 cm
Height (English) 15 in.
Height (Metric) 38 cm
Weight (English) 62 lb.
Weight (Metric) 28 kg
Width (English) 26.8 in.
Width (Metric) 68 cm
Total Service Coverage 1 year manufacturer warranty only
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.