missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Flex Purification System, SmartStart, 96 PCR head

Catalog No. A39967
Encompass
Change view
Click to view available options
Quantity:
1 package
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39967 1 package
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39967 Supplier Thermo Scientific™ Supplier No. A39967

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prepare samples efficiently from a variety of materials with the Thermo Scientific KingFisher Flex Purification System which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, or cell purification from virtually any source. Package includes the KingFisher Flex Purification System and SmartStart Orientation.

  • Efficiently prepare samples from a variety of materials
  • System offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, or cell purification from virtually any source
  • Fully automated system yields high-speed purification of nucleic acids, proteins, and cells
  • High-throughput system processes up to 96 samples in less than 20 minutes
  • Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands
  • Package includes the KingFisher Flex Purification System and SmartStart Orientation
  • Easy-to-use BindIt Software provides instrument control, protocol creation, and modification
  • Ready-made protocols available for different type of applications

Get up and running fast with SmartStart Orientation

  • The SmartStart Orientation included in this package is performed by a Field Application Scientist (FAS) in your lab and helps to - 
    • Get up and running quickly
    • Get connected to your local network and Cloud
    • Choose appropriate reagents and consumables to meet your needs
  • Content will vary depending on customer's level of knowledge and experience

Specifications

Volume (Metric) Processing 20 to 100 μL
Plates per Deck 24 or 96
Format 96-well Plate
Description KingFisher™ Flex Purification System
For Use With (Application) Genomics and Proteomics, Target Identification, Veterinary Assays, Biomarker Discovery, Quality Control, Forensics
For Use With (Equipment) Nucleic Acid Purification Systems
Includes KingFisher™ Flex Purification System, 96 PCR head and SmartStart 1 Day Training
Particle Size (Metric) ca. >1 μm
Quantity 1 package
Customer Education 1 day at customer site
Total Service Coverage 1 year manufacturer warranty only
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.