Learn More
Thermo Scientific™ KingFisher™ Duo Prime Purification System with SmartStart
Catalog No. A39956PKG
Change view
Click to view available options
Quantity:
1 package
Catalog No. Quantity
A39956PKG 1 package
1 options
Catalog No. A39956PKG Supplier Thermo Scientific™ Supplier No. A39956
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reap high yields of quality nucleic acids from a variety of starting materials with the Thermo Scientific KingFisher Duo Prime Purification System. Package includes the KingFisher Duo Prime Purification System and SmartSmart Orientation.
  • Reap high yields of quality nucleic acids from a variety of starting materials including cell-free body fluids, blood, bacteria, cell culture, tissue, and plant samples
  • This compact instrument is ideal for laboratories requiring nucleic acid purification and is easily modified for use in proteomics and cell isolation
  • It provides excellent reproducibility with minimal hands-on time and has a small footprint in the lab
  • A UV lamp for easy decontamination and optional bar-code reader enhance its functionality
  • Automated magnetic bead technology allows excellent recovery of samples with great reproducibility and efficiency
  • Optional barcode reader allows samples to be tracked in the internal software
  • Package includes the KingFisher Duo Prime Purification System and SmartStart Orientation
  • Optimized for use with KingFisher Pure DNA and RNA Isolation Kits and Protocols
Get up and running fast with SmartStart Orientation
  • The SmartStart Orientation included in this package is performed by a Field Application Scientist (FAS) in the lab and helps to - 
    • Get up and running quickly
    • Get connected to your local network and Cloud
    • Choose appropriate reagents and consumables to meet specific needs
  • Content will vary depending on customer's level of knowledge and experience
Specifications
Volume (Metric) Processing 96 DW plate 50 to 1000 μL, 24 DW plate 200 to 5000 μL, Elution strip 30 to 130 μL
Description KingFisher™ Duo Prime Purification System
For Use With (Application) DNA and RNA Isolation From Various Starting Materials, Proteomic Applications, Cell Isolation
For Use With (Equipment) Nucleic Acid Purification Systems
Includes KingFisher™ Duo Prime Purification System and SmartStart 1 Day Training
Particle Size (Metric) ca. >1 μm
Quantity 1 package
Customer Education 1 day at customer site
Total Service Coverage 1 year manufacturer warranty only
Where can I find the manual for the BindIt Software?
The User Manual for the BindIt Software can be downloaded from the BindIt Software product page online, under the Documents & Downloads section. It is also included within the BindIt Software installation file under "Documentation".
Do you have data that shows how the UV lamp of the KingFisher Duo Prime instrument gets rid of bacterial contamination?
Yes we have an application note that demonstrates the lethal effect of UVC irradiation on bacterial growth and deactivation of human gDNA, "Decontamination of the KingFisher Duo Prime processing chamber using UV light".
What type of tip heads do I receive when I purchase the KingFisher Duo Prime instrument?
You will receive the following tip heads when you purchase a KingFisher Duo Prime instrument:
• 6-tip head (for 24 deep-well plate)
• 12-tip head (for 96 deep-well plate)
Do you sell elution strips for the KingFisher Duo Prime 24 Deep-Well (6-tip) format?
We do not sell elution strips for the KingFisher Duo Prime 24 Deep-Well format. A 24 Deep-Well plate must be used for elution with the 24 Deep-Well protocol.
Are the Matrix 500 microliter ScrewTop Tubes (Cat. No. 3743AMB) compatible with the Kingfisher Duo Prime barcode reader?
Yes, the KingFisher Duo Prime barcode reader is able to read the 2D barcode on the Matrix 500 µL ScrewTop Tubes. The barcode reader can generate an error if the tubes are in the tube rack and the barcode position is not able to be scanned. If this occurs, we recommend scanning each tube individually.
Can I run two protocols simultaneously on the KingFisher Duo Prime?
You can only run two protocols on the KingFisher Duo Prime if the following criteria are met:
• Both scripts only use one plate
• Both scripts are using the 96-well format
The 24-well plate format always requires two plates, so it cannot run two protocols. If each script uses one plate and it is the 96-well format, you can run two protocols using the following instructions:
1. Start the first protocol as normal.
2. Once the first protocol has started, scroll back over to the protocol selection tab and select the second script. The second script can be the same as the first script or a different script.
3. Start the second protocol.
4. Select "Start" and load the second plate.
The instrument will be processing the first plate while you start the second plate. When the first plate is done, the KingFisher Duo Prime instrument will automatically start processing the second plate.
How do I use BindIt software 4.0 to load a script from my computer to the KingFisher Flex or KingFisher Duo/Duo Prime instrument?
Please follow these steps to load a script from your computer to the KingFisher Flex or KingFisher Duo/Duo Prime instrument:
1. Use the USB cable to connect the PC to the instrument.
2. Turn on the instrument.
3. Open the BindIt software and click the Connect button.
4. Select "Find New Instrument." BindIt will automatically connect to the instrument and the instrument screen will say "Computer Control".
5. Click the "Transfer" Button.
6. Click the "Upload" button.
7. Browse to find the file and double-click it.
8. In the new window that opens:
a. KingFisher Flex: you will be prompted to enter a new name – this is not necessary if the name only contains letters, digits, dash or underscore.
• If the name contains spaces, apostrophes or any other character, you should enter a new name containing only letters, digits, dash (-) or underscore (_).
• Click the "Category" pulldown to select the instrument folder you want to load the script to.
• Click OK.
b. KingFisher Duo/Duo Prime: The KingFisher Duo and Duo Prime instruments accept all characters in the filename.
• Click the "Category" pulldown to select the instrument folder you want to load the script to.
• Click OK.
Do the KingFisher Flex and KingFisher Duo instruments come with pre-loaded scripts?
The KingFisher Flex and KingFisher Duo instruments no longer come with pre-installed scripts. If your instrument begins with the following serial numbers, it will not have pre-loaded scripts:
• KingFisher Flex serial number 711-1951 or greater
• KingFisher Duo Prime serial number 706-1366 or greater
Do I receive training when I purchase the KingFisher Flex or KingFisher Duo Prime instruments?
No training is provided with the purchase of a KingFisher Flex or KingFisher Duo Prime instrument, but you can purchase training separately. We offer onsite half-day and full-day trainings for the KingFisher Flex and KingFisher Duo Prime instruments, depending on your needs. Please contact your local instrument sales representative to obtain a quote. The training will be led by your regional Field Application Scientist.
Where can I find a script for processing a large volume of blood using the MagMAX Multi-Sample Ultra kit (Cat. No. A25597) with the KingFisher Flex or KingFisher Duo Prime instruments?
Scripts for processing large volumes of whole blood (up to 1 mL) can be found on the MagMAX Multi-Sample Ultra kit product page under the Product Literature section at the bottom of the page. There are two scripts available:
• KingFisher DUO file: A25597_Large_Volume_Blood.bdz
• KingFisher Flex file: A25597_Large_Volume_Blood.bdz
*Use the page scrolling function to see the complete list of product literature for this product.
What kind of consumables do I need for the KingFisher Duo Purification System?
The KingFisher Duo Purification System consumables are listed on the following website and they vary depending on whether you are using the 6- or 12-pin magnet: https://www.thermofisher.com/order/catalog/product/97003500
For Research Use Only. Not for use in diagnostic procedures.