missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format

Catalog No. 97002570
Encompass
Change view
Click to view available options
Product Type:
Barcoded Tip Comb
Combi Pack
Elution Strip
Elution Strip Cap
Tip Comb
For Use With (Equipment):
KingFisher Apex
KingFisher Duo Prime only
KingFisher Flex and Presto
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
97002570 Tip Comb KingFisher Apex
22387029 Tip Comb KingFisher Flex and Presto
14387291 Tip Comb KingFisher Duo Prime only
14387293 Elution Strip KingFisher Duo Prime only
14387737 Elution Strip Cap KingFisher Duo Prime only
14387294 Combi Pack KingFisher Duo Prime only
14387354 Barcoded Tip Comb KingFisher Apex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 97002570 Supplier Thermo Scientific™ Supplier No. 97002570

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Apex, KingFisher Flex, Duo Prime, and Presto instruments.

Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.

Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications

Product Type Tip Comb
Format 96 DW Comb
Capacity 96 Samples
Description KingFisher Apex 96 Combi tip comb (for Apex only)
Disposable Single-use
For Use With (Equipment) KingFisher Apex
Product Line KingFisher
Quantity 100/Pk.
Do you offer an alternative for the discontinued Pharma MagMAX Express-96 Deep-Well Tip Combs (Cat. No. A31537)?

Yes, we recommend using KingFisher 96 tip combs for deep-well format (Cat. No. 97002534 or 97002570).

Are KingFisher Plastics for 96-deep well format autoclavable?

No, KingFisher Plastics for 96-deep well format are not autoclavable. They won't melt, but they will get deformed.

Can I reuse the Kingfisher Plastics for 96 deep-well format?

The Kingfisher Plastics for 96 deep-well format are recommended for one-time use only, and we don't recommend reusing these products.

Are the KingFisher Plastics for 96 deep-well format CE marked?

The KingFisher Plastics for 96 deep-well format are not CE marked.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.