Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
| Product Type | Combi Pack |
| Format | 96 DW Comb |
| Description | KingFisher combi pack for microtiter 96 deep-well plate (includes 8 plates, 8 tip combs, and 8 elution strips and caps) (for Duo Prime only). |
| For Use With (Equipment) | KingFisher Duo Prime only |
| Includes | 8 plates, 8 tip combs, and 8 elution strips and caps |
| Product Line | KingFisher |
Frequently Asked Questions (FAQs)
Yes, we recommend using KingFisher 96 tip combs for deep-well format (Cat. No. 97002534 or 97002570).
No, KingFisher Plastics for 96-deep well format are not autoclavable. They won't melt, but they will get deformed.
The Kingfisher Plastics for 96 deep-well format are recommended for one-time use only, and we don't recommend reusing these products.
The KingFisher Plastics for 96 deep-well format are not CE marked.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.