missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Apex Dx Magnetic Heads

Catalog No. B24079930
Change view
Click to view available options
Format:
Apex Dx 24 Combi Head
Apex Dx 96 Combi Head
Apex Dx 96 Deep Well Head
Apex Dx 96 PCR Head
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
B24079930 Apex Dx 96 Deep Well Head
B24079940 Apex Dx 24 Combi Head
B24079920 Apex Dx 96 Combi Head
B24079910 Apex Dx 96 PCR Head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. B24079930 Supplier Thermo Scientific™ Supplier No. B24079930
Only null left

The Thermo Scientific KingFisher Apex Dx Purification Instrument allows for extraction of nucleic acids from human specimens with a small, compact footprint making it ideal for use in diagnostic and development workflows.

KingFisher Apex Dx Magnetic Heads are for use with the KingFisher Apex Dx Purification Instrument. The instrument accommodates two interchangeable magnetic heads (it comes with one included). The interchangeable heads and heating blocks provide flexibility in the extraction of nucleic acids, proteins, and cells with a small, compact footprint, making the instrument ideal for use in diagnostic and development workflows.

With its intuitive software, automation capabilities, and revolutionary magnetic particle separation technology, the KingFisher Apex Dx Purification Instrument provides high quality, exceptional reliability, and outstanding run-to-run reproducibility for clinicians and assay developers seeking an IVD sample preparation solution for their diagnostic workflow.

Specifications

Product Type Nucleic Acid Extraction and Cell Separation Instrument
Sample Type Viral Samples
Format Apex Dx 96 Deep Well Head
Disposable Reusable
Quantity 1 head

For in vitro diagnostic use.