missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Kinetin | |
| C10H9N5O | |
| 1 g | |
| QANMHLXAZMSUEX-UHFFFAOYSA-N | |
| N-(furan-2-ylmethyl)-7H-purin-6-amine | |
| 3830 | |
| 215.22 | |
| Crystalline Powder |
| 525-79-1 | |
| MFCD00075757 | |
| kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl | |
| C1=COC(=C1)CNC2=NC=NC3=C2NC=N3 | |
| 215.216 | |
| CHEBI:27407 | |
| ≥99.0% (HPLC,T) |
Chemical Identifiers
| 525-79-1 | |
| 215.216 | |
| QANMHLXAZMSUEX-UHFFFAOYSA-N | |
| 3830 | |
| N-(furan-2-ylmethyl)-7H-purin-6-amine |
| C10H9N5O | |
| MFCD00075757 | |
| kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl | |
| CHEBI:27407 | |
| C1=COC(=C1)CNC2=NC=NC3=C2NC=N3 |
Safety and Handling
RTECSNumber : AU6270000
TSCA : Yes