missing translation for 'onlineSavingsMsg'
Learn More

Kinetin 99.0+%, TCI America™

Catalog No. p-7131739
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00091G 1 g
K00095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00091G Supplier TCI America Supplier No. K00091G
Only null left

Chemical Identifiers

CAS 525-79-1
Molecular Formula C10H9N5O
Molecular Weight (g/mol) 215.216
MDL Number MFCD00075757
InChI Key QANMHLXAZMSUEX-UHFFFAOYSA-N
Synonym kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
PubChem CID 3830
ChEBI CHEBI:27407
IUPAC Name N-(furan-2-ylmethyl)-7H-purin-6-amine
SMILES C1=COC(=C1)CNC2=NC=NC3=C2NC=N3

Specifications

Quantity 1 g
Formula Weight 215.22
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Kinetin
RTECSNumber AU6270000
TSCA Yes